Learn More
Biolegend Purified anti-ALDH1L1
Supplier:  Biolegend 856801
Encompass_Preferred
Purified anti-ALDH1L1 [N103/39]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, ICC, IHC-F; Size: 25 ug
Catalog No. 50-207-4777