Learn More
Biolegend Purified anti-β-Amyloid, aggregated
Supplier:  Biolegend 871202
Encompass_Preferred
Purified anti-β-Amyloid, aggregated [A18142A]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: IHC-P, WB; Size: 100 ug
Catalog No. 50-207-5076