Learn More
Biolegend Purified anti-AND-1 (WDHD1)
Supplier:  Biolegend 630302
Encompass_Preferred
Purified anti-AND-1 (WDHD1) [20G10]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse; Apps: WB, IHC-P; Size: 100 ug
Catalog No. 50-170-845