Learn More
Biolegend Purified anti-ATG17
Supplier:  Biolegend 620002
Encompass_Preferred
Purified anti-ATG17 [W16188A]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: WB; Size: 100 ug
Catalog No. 50-208-6772