Learn More
Biolegend Purified anti-Apo-E
Supplier:  Biolegend 674802
Encompass_Preferred
Purified anti-Apo-E [3B3C32]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: WB; Size: 100 ug
Catalog No. 50-208-7336