Learn More
Biolegend Purified anti-Arginase 1
Supplier:  Biolegend 678801
Encompass_Preferred
Purified anti-Arginase 1 [O94E6/ARG1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, KO/KD-WB; Size: 25 ug
Catalog No. 50-208-7394