Learn More
Biolegend Purified anti-Ataxin-3
Supplier:  Biolegend 650402
Encompass_Preferred
Purified anti-Ataxin-3 [1H9-2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: WB; Size: 100 ug
Catalog No. 50-171-071