Learn More
Biolegend Purified anti-β-actin
Supplier:  Biolegend 643801
Encompass_Preferred
Purified anti-β-actin [2F1-1]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse, Rat; Apps: WB, ICC; Size: 25 ug
Catalog No. 50-170-956