Learn More
Biolegend Purified (azide-free) anti-β-Amyloid, 1-16
Supplier:  Biolegend 803003
Encompass_Preferred
Purified (azide-free) anti-β-Amyloid, 1-16 [6E10]; Isotype: Mouse IgG1; Reactivity: Human; Apps: WB, Direct ELISA, IHC-P, IHC-F, EM; Size: 1 mL
Catalog No. 50-102-9535