Learn More
Biolegend Purified (azide-free) anti-β-Amyloid, 17-24
Supplier:  Biolegend 800701
Encompass_Preferred
Purified (azide-free) anti-β-Amyloid, 17-24 [4G8]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: IHC-P, Direct ELISA, IHC-F, ICC, IP; Size: 200 uL
Catalog No. 50-102-9492