Learn More
Biolegend Purified anti-Bcl-2
Supplier:  Biolegend 633501
Encompass_Preferred
Purified anti-Bcl-2 [BCL/10C4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: WB, IP, ICC, IHC-F; Size: 25 ug
Catalog No. 50-170-872