Learn More
Biolegend Purified anti-mouse/human Bcl-6
Supplier:  Biolegend 648301
Encompass_Preferred
Purified anti-mouse/human Bcl-6 [IG191E/A8]; Isotype: Mouse IgG1; Reactivity: Human, Mouse; Apps: WB, ChIP, IHC-P, IP; Size: 25 ug
Catalog No. 50-171-037