Learn More
Biolegend Purified anti-BTK Phospho (Tyr551)/ITK Phospho (Tyr512)
Supplier:  Biolegend 646902
Encompass_Preferred
Purified anti-BTK Phospho (Tyr551)/ITK Phospho (Tyr512) [A16064A]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, ICFC; Size: 100 ug
Catalog No. 50-208-6948