Learn More
Biolegend Purified anti-BTK Phospho (Tyr223)
Supplier:  Biolegend 601702
Encompass_Preferred
Purified anti-BTK Phospho (Tyr223) [A16128B]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse; Apps: WB, ICFC; Size: 100 ug
Catalog No. 50-208-6631