Learn More
Biolegend Purified anti-C/EBP β (2 isoforms C/EBP β, LAP)
Supplier:  Biolegend 626002
Encompass_Preferred
Purified anti-C/EBP β (2 isoforms C/EBP β, LAP) [A16]; Isotype: Mouse IgG; Reactivity: Mouse (recognizes 35 and 32 kD LAP forms); Apps: WB; Size: 100 ug
Catalog No. 50-170-780