Learn More
Biolegend Purified anti-Caspase-1
Supplier:  Biolegend 645101
Encompass_Preferred
Purified anti-Caspase-1 [5B10]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: WB, IHC; Size: 25 ug
Catalog No. 50-170-989