Learn More
Biolegend Purified anti-Caspase 3
Supplier:  Biolegend 634101
Encompass_Preferred
Purified anti-Caspase 3 [Poly6341]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human; Apps: WB, IP; Size: 125 uL
Catalog No. 50-170-890