Learn More
Biolegend Purified anti-CCL11
Supplier:  Biolegend 683202
Encompass_Preferred
Purified anti-CCL11 [L403H11]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Capture; Size: 500 ug
Catalog No. 50-208-7466