Learn More
Biolegend Purified anti-CD55
Supplier:  Biolegend 868802
Encompass_Preferred
Purified anti-CD55 [M033]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IHC-P; Size: 100 ug
Catalog No. 50-207-5026