Learn More
Biolegend Purified anti-CDC2 Phospho (Tyr15) [A21009D]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICFC; Size: 100 ug
Supplier:  Biolegend 613852
Encompass_Preferred
Purified anti-CDC2 Phospho (Tyr15) [A21009D]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICFC; Size: 100 ug
Catalog No. 50-237-8584