Learn More
Biolegend Purified anti-CDK4
Supplier:  Biolegend 690802
Encompass_Preferred
Purified anti-CDK4 [W15200A]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: WB; Size: 100 ug
Catalog No. 50-208-7588