Learn More
Biolegend Purified anti-Chk2
Supplier:  Biolegend 686602
Encompass_Preferred
Purified anti-Chk2 [P81C12B8]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: WB, ICC, KO/KD-WB; Size: 100 ug
Catalog No. 50-208-7533