Learn More
Biolegend Purified anti-Claudin-1
Supplier:  Biolegend 697801
Encompass_Preferred
Purified anti-Claudin-1 [9E12C72]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB; Size: 25 ug
Catalog No. 50-208-7705