Learn More
Biolegend Purified anti-complement C3/C3b/iC3b
Supplier:  Biolegend 846402
Encompass_Preferred
Purified anti-complement C3/C3b/iC3b [7C12/C3b]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, FC, Direct ELISA, ELISA Detection, RIA; Size: 100 ug
Catalog No. 50-207-4497