Learn More
Biolegend Purified anti-Connexin 43, 1-20
Supplier:  Biolegend 849901
Encompass_Preferred
Purified anti-Connexin 43, 1-20 [P1E11]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: WB; Size: 25 ug
Catalog No. 50-207-4580