Learn More
Biolegend Purified anti-Cyclin A
Supplier:  Biolegend 644001
Encompass_Preferred
Purified anti-Cyclin A [E23.1]; Isotype: Mouse IgG2a; Reactivity: Human, Mouse, Bovine; Apps: WB, ICFC; Size: 25 ug
Catalog No. 50-170-959