Learn More
Biolegend Purified anti-Cyclin B1
Supplier:  Biolegend 647901
Encompass_Preferred
Purified anti-Cyclin B1 [V152]; Isotype: Mouse IgG1; Reactivity: Human, Cross-Reactivity: Mouse; Apps: WB, IF, IHC; Size: 25 ug
Catalog No. 50-171-028