Learn More
Biolegend Purified anti-Cyclin D3
Supplier:  Biolegend 684902
Encompass_Preferred
Purified anti-Cyclin D3 [DCS-22]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: WB, ICFC, KO/KD-WB; Size: 100 ug
Catalog No. 50-208-7496