Learn More
Biolegend Purified anti-Cytokeratin 18
Supplier:  Biolegend 689102
Encompass_Preferred
Purified anti-Cytokeratin 18 [LDK18]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICC, IHC-P; Size: 100 ug
Catalog No. 50-208-7564