Learn More
Biolegend Purified anti-Cytokeratin 18
Supplier:  Biolegend 628401
Encompass_Preferred
Purified anti-Cytokeratin 18 [DA-7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICC, ELISA, IHC-P, IP; Size: 25 ug
Catalog No. 50-170-815