Learn More
Biolegend Purified anti-Cytokeratin 6
Supplier:  Biolegend 606101
Encompass_Preferred
Purified anti-Cytokeratin 6 [W17006B]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB, ICC; Size: 25 ug
Catalog No. 50-208-6645