Learn More
Biolegend Purified anti-DDX17 (p82)
Supplier:  Biolegend 657202
Encompass_Preferred
Purified anti-DDX17 (p82) [1B5B09]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, IP, ICC; Size: 100 ug
Catalog No. 50-208-7126