Learn More
Biolegend Purified anti-EGFR
Supplier:  Biolegend 617501
Encompass_Preferred
Purified anti-EGFR [A18011A]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICC, IP, ICFC; Size: 25 ug
Catalog No. 50-208-6752