Learn More
Biolegend Purified anti-eIF3I (p36)
Supplier:  Biolegend 646701
Encompass_Preferred
Purified anti-eIF3I (p36) [17C11]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: WB; Size: 25 ug
Catalog No. 50-171-011