Learn More
Biolegend Purified anti-ERp57
Supplier:  Biolegend 937301
Encompass_Preferred
Purified anti-ERp57 [W19080B]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB; Size: 25 ug
Catalog No. 50-207-5368