Learn More
Biolegend Purified anti-FBXL5
Supplier:  Biolegend 672703
Encompass_Preferred
Purified anti-FBXL5 [3F5G12G9]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: WB; Size: 25 ug
Catalog No. 50-208-7307