Learn More
Biolegend Purified anti-FIG4/SAC3
Supplier:  Biolegend 861801
Encompass_Preferred
Purified anti-FIG4/SAC3 [N202/7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: WB; Size: 25 ug
Catalog No. 50-207-4855