Learn More
Biolegend Purified anti-Flotillin-1
Supplier:  Biolegend 849802
Encompass_Preferred
Purified anti-Flotillin-1 [W16108A]; Isotype: Rat IgG2a, κ; Reactivity: Human, Rat, Mouse; Apps: WB, ICC; Size: 100 ug
Catalog No. 50-207-4577