Learn More
Biolegend Purified anti-FOXA1
Supplier:  Biolegend 612001
Encompass_Preferred
Purified anti-FOXA1 [2F83]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICC, IHC-P; Size: 25 ug
Catalog No. 50-208-6706