Learn More
Biolegend Purified anti-Fyn
Supplier:  Biolegend 626501
Encompass_Preferred
Purified anti-Fyn [FYN-59]; Isotype: Mouse IgG; Reactivity: Human, Mouse; Apps: WB; Size: 25 ug
Catalog No. 50-170-784