Learn More
Biolegend Purified anti-Gelsolin
Supplier:  Biolegend 623001
Encompass_Preferred
Purified anti-Gelsolin [W18073A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB, IP; Size: 25 ug
Catalog No. 50-208-6810