Learn More
Biolegend Purified anti-GFAP
Supplier:  Biolegend 801103
Encompass_Preferred
Purified anti-GFAP [MCA-5C10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB, ICC, IHC-F; Size: 25 ug
Catalog No. 50-208-9834