Learn More
Biolegend Purified anti-GFP
Supplier:  Biolegend 668206
Encompass_Preferred
Purified anti-GFP [1GFP63]; Isotype: Mouse IgG2a; Reactivity: Aequorea victoria; Apps: WB; Size: 25 ug
Catalog No. 50-208-7265