Learn More
Biolegend Purified anti-H2A.X Phospho (Ser139)
Supplier:  Biolegend 613401
Encompass_Preferred
Purified anti-H2A.X Phospho (Ser139) [2F3]; Isotype: Mouse IgG1, κ; Reactivity: Human, reacts with Ser139-phosphorylated H2A.X; Apps: WB, ICC, ICFC; Size: 25 ug
Catalog No. 50-170-651