Learn More
Biolegend Purified anti-HCN4
Supplier:  Biolegend 857002
Encompass_Preferred
Purified anti-HCN4 [N114/10]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: WB, IHC-P; Size: 100 ug
Catalog No. 50-207-4786