Learn More
Biolegend Purified anti-HDAC2
Supplier:  Biolegend 680102
Encompass_Preferred
Purified anti-HDAC2 [13G8C67]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, IP, ICC, KO/KD-WB; Size: 100 ug
Catalog No. 50-208-7408