Learn More
Biolegend Purified anti-HDAC5
Supplier:  Biolegend 693401
Encompass_Preferred
Purified anti-HDAC5 [W15202A]; Isotype: Rat IgG2b, κ; Reactivity: Human, Mouse; Apps: WB; Size: 25 ug
Catalog No. 50-208-7623