Learn More
Biolegend Purified anti-HDAC8
Supplier:  Biolegend 685502
Encompass_Preferred
Purified anti-HDAC8 [W15090A]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, KO/KD-WB, ICC; Size: 100 ug
Catalog No. 50-208-7506