Learn More
Biolegend Purified anti-Histone H2B
Supplier:  Biolegend 606301
Encompass_Preferred
Purified anti-Histone H2B [W17025A]; Isotype: Rat IgG2a, λ; Reactivity: Human, Mouse, Drosophila; Apps: WB, IHC-P; Size: 25 ug
Catalog No. 50-208-6647