Learn More
Biolegend Purified anti-Histone H3 Phospho (Ser10)
Supplier:  Biolegend 650801
Encompass_Preferred
Purified anti-Histone H3 Phospho (Ser10) [11D8]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: WB; Size: 25 ug
Catalog No. 50-171-078